Swabs with transport medium
Catalog # LPIT118898
Supplier: LP ITALIANA
Specifications
- Popis:Výtěry s transportním médiem
- Shaft/head:Plastové/hedvábí
- Transport medium:Stuart gel
- Bal.:250
Specifications
About this item
Swabs with transport medium.
Specially designed for common applications.
Packaging: Individually wrapped in a peel-pouch.
Caution: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904