Orders LinkContactUsLinkComponent
Scraper, Ayre
Scraper, Ayre
Ayre scraper, width 18 mm, 180 mm
Catalog # LPIT133012
Supplier:   LP ITALIANA
Scraper, Ayre
Catalog # LPIT133012
Supplier:  LP ITALIANA
Scraper, Ayre
Scraper, Ayre
Ayre scraper, width 18 mm, 180 mm
Price
2 740,00 Kč
Log in for availability
1 * 1.000 KS
Product Details & Documents
PS, white.
Caution: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904
Specifications
  • Popis:
    Ayre scraper, width 18 mm
  • Délka:
    180 mm