Orders LinkContactUsLinkComponent
Bandages, Askina® Cel
Bandages, Askina® Cel
Cellulose dressing, 100% bleached cellulose, roll (36 cm wide), 1000 g
Katalogové číslo 115-2515
Dodavatel:   B. Braun
Bandages, Askina® Cel
Katalogové číslo 115-2515
Dodavatel:  B. Braun
Bandages, Askina® Cel
Bandages, Askina® Cel
Cellulose dressing, 100% bleached cellulose, roll (36 cm wide), 1000 g
Přejít na:
Price
660,00 Kč
Přihlásit se pro dostupnost
1 * 1 kg
Product Details & Documents
Highly absorbent dressing made of 100% chlorine-free bleached cellulose.
  • Highly bleached
  • Price-effective absorbent material
Specifikace
  • Popis:
    Cellulose dressing, 100% bleached cellulose, roll (36 cm wide), 1000 g