Orders LinkContactUsLinkComponent
Automatic burettes, Pellet pattern, class B
Automatic burettes, Pellet pattern, class B
50 ml, 0,1 ml, 0,08 ± ml
Katalogové číslo WITG3.114.506
Dodavatel:   witeg Labortechnik
Automatic burettes, Pellet pattern, class B
Katalogové číslo WITG3.114.506
Dodavatel:  witeg Labortechnik
Automatic burettes, Pellet pattern, class B
Automatic burettes, Pellet pattern, class B
50 ml, 0,1 ml, 0,08 ± ml
Price
12 080,00 Kč
Přihlásit se pro dostupnost
1 * 2 KS
Product Details & Documents
Automatic burettes, Pellet pattern, class B, made of amber borosilicate glass, short line graduation.
  • Stopcock, with hollow NS glass key, between burette and reservoir
  • Lateral stopcock, with solid NS glass key
  • Complete with 2000 ml reservoir bottle, made of DURAN® borosilicate glass, without aspirator bulb
Specifikace
  • Kapacita:
    50 ml
  • Dělení:
    0,1 ml
  • Tolerance:
    0,08 ± ml
Accessories
Informace o produktu
  • Katalogové číslo
    Kapacita
    Dostupnost
    Price
  • Reservoir bottle, amber, neck with NS 29/32 joint, 2000 ml
    Reservoir bottle, amber, neck with NS 29/32 joint, 2000 ml
    SCOT211596369
    2000 ml
    Přihlásit se pro dostupnost
    1 * 1 KS - 2 920,00 Kč
Informace o produktu
  • Katalogové číslo
    Kapacita
    Dostupnost
    Price
  • Reservoir bottle, clear, neck with NS 29/32 joint, 2000 ml
    Reservoir bottle, clear, neck with NS 29/32 joint, 2000 ml
    SCOT211596303
    2000 ml
    Přihlásit se pro dostupnost
    1 * 1 KS - 2 110,00 Kč