Urine containers, with screw cap and spout
Catalog # LPIT108039
Supplier: LP ITALIANA
Specifications
- Kapacita:2500 ml
- Š×H×V:255×100×100 mm
- Bal.:16
Specifications
About this item
Container made of HDPE.
- Good chemical resistance
- Good temperature resistance