Orders LinkContactUsLinkComponent
Separating funnels, cylindrical
Separating funnels, cylindrical
100 ml, 19/26 NS
Katalogové číslo WITG2.643.100
Dodavatel:   witeg Labortechnik
Separating funnels, cylindrical
Katalogové číslo WITG2.643.100
Dodavatel:  witeg Labortechnik
Separating funnels, cylindrical
Separating funnels, cylindrical
100 ml, 19/26 NS
Přejít na:
Price
1 710,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Cylindrical separatory funnels, made of borosilicat glass, with Witaflo PTFE needle valve stopcock.
  • Graduated
  • Very good chemical resistance
  • Very good temperature resistance
Specifikace
  • Velikost objímky:
    19/26 NS
  • Kapacita:
    100 ml