Orders LinkContactUsLinkComponent
Multi-level removal funnels, Squibb
4000 ml, 45/40 NS
Katalogové číslo WITG2.636.004
Dodavatel:   witeg Labortechnik
Multi-level removal funnels, Squibb
Katalogové číslo WITG2.636.004
Dodavatel:  witeg Labortechnik
Multi-level removal funnels, Squibb
Multi-level removal funnels, Squibb
4000 ml, 45/40 NS
Přejít na:
Price
13 410,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Multi-level removal funnel, Squibb, made of borosilicate glass 3.3, with PTFE-outlet valve and plastic stopper.
  • Easy to read graduation
  • Very good chemical resistance
  • Very good temperature resistance
Specifikace
  • Velikost objímky:
    45/40 NS
  • Kapacita:
    4000 ml