Orders LinkContactUsLinkComponent
Separating funnels, spherical
Separating funnels, spherical
50 ml, 19/26 NS
Katalogové číslo WITG2.620.050
Dodavatel:   witeg Labortechnik
Separating funnels, spherical
Katalogové číslo WITG2.620.050
Dodavatel:  witeg Labortechnik
Separating funnels, spherical
Separating funnels, spherical
50 ml, 19/26 NS
Přejít na:
Price
3 290,00 Kč
Přihlásit se pro dostupnost
1 * 2 KS
Product Details & Documents
Separatory funnel, globe (ball) shaped, made of borosilicate glass 3.3, with detachable PTFE-stopcock and PE stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Specifikace
  • Velikost objímky:
    19/26 NS
  • Kapacita:
    50 ml