Bends
19/26 + 24/29 NS
Katalogové číslo WITG1.000.004
Dodavatel: witeg Labortechnik
Přejít na:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Inclined at an angle of 90°. With 2 standard ground cones.
Specifikace
- Rozměry zábrusu:19/26 + 24/29 NS