Volumetric flasks, class A, trapezoidal shape, blue markings
Katalogové číslo HECH42110024
Dodavatel: GLASWARENFABRIK KARL HECHT
Specifikace
- NZ:7/16
- Kapacita:10 ml
- Bal.:1
- Tolerance:0,025 ± ml
Specifikace
O tomto produktu
DURAN® Borosilicate glass 3.3 with PP stopper.
- Calibrated to contain (TC, In)
- Blue markings
- Conformity certified (DE-M marking)
The base is considerably larger than of standard volumetric flasks with same volume, thus improved stability.
In accordance with DIN EN ISO 1042