Horseradish peroxidase
Dodavatel: Merck
Certifikáty
O tomto produktu
Specifikace
- Čí. CAS:9003-99-0
- Druhy:Horseradish
- Podmínky uskladnění:Freezer
- MDL čí.:MFCD00071339
- Název enzymu:Peroxidase
Specifikace
Frequently Bought Together
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Dodavatel: Dr. Weigert
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Diethylether ≥99.5%, AR®, Macron Fine Chemicals™
Dodavatel: MACRON AVANTOR BRAND
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Aceton ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Sebakoyldichlorid, Sigma-Aldrich®
Dodavatel: Merck
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, Sigma-Aldrich®
Dodavatel: Merck