ContactUsLinkComponent
Phosphoramidon
Phosphoramidon
  786-054
 :  
Phosphoramidon
  786-054
 :  
  • Bal.:
    10 mg
  • Čí. CAS:
    36357-77-4
  • Název proteinu/peptidu:
    Phosphoramidon
Phosphoramidon inhibits some metalloproteases, including thermolysin, collagenase and bacterial metallo proteases from Bacillus subtilis, Streptomyces griseus and Pseudomonas aeruginosa (metallo elastase). Soluble in water, methanol and DMSO.