Phenylmercury nitrate (basic), Sigma-Aldrich®
Catalog # SIAL79135-50G
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:50 g
- Sekvence:3368
Specifications
About this item
:
UN:1895
ADR:6.1,II
CAS Number:8003-05-2
Formula:C₁₂H₁₁Hg₂NO₄