Microfiltration filters, MiniKap®
Catalog # SPECMK2M204V6N
Supplier: Spectrum Laboratories
Specifications
- Filtrační plocha:225 cm²
- Velikost pórů:0,2 µm
- Type:Microfiltration filters, MiniKap®
- Vstup/výstup:MNPT 1/4 inches / MNPT 1/4 inches
- Objem vzorku:500 - 1000 ml
- Typ:Microfiltration filters, MiniKap®
- Bal.:6
Specifications
About this item
These Point-of-use microfiltration filters are excellent for water and gas filtration systems. MiniKap® HF filter provides the filtration efficiency of the 0,2 µm DynaFibre® and air vent fibre in a device conveniently equipped with a ¼'' MNPT, Luer-Lok or hose barb inlet connection.