Stopcock for aspirator bottles
SCOT241470409
- Popis:Stopcock for aspirator bottles, PTFE, for GL 32 screw thread, hole Ø 8 mm
- Sekvence:02
- Bal.:1
Borosilicate glass 3.3, clear, with PTFE spindle, for GL 32 screw thread.
Borosilicate glass 3.3, clear, with PTFE spindle, for GL 32 screw thread.