Filtry stříkačkové, Puradisc™ Aqua 30, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
About this item
Puradisc™ Aqua 30 syringe filters are specifically designed for environmental sample filtration prior to dissolved organic carbon (DOC) and chemical oxygen demand (COD) analysis.
- From the Puradisc FP 30 range
- Low protein binding, hydrophilic cellulose acetate syringe filter media
- Prewashed cellulose acetate (CA) syringe filter for reduced organic carbon extractables
- Designed for sample preparation in support of dissolved organic carbon and chemical oxygen demand analysis
- Standard inlet and outlet luer connectors
DOC (dissolved organic carbon) and COD (chemical oxygen demand) analysis are common methods to determine organic pollutant levels.
Precisely filtering a sample is a crucial step in DOC and COD, and the quality of the syringe filter should preserve sample integrity. Puradisc™ Aqua 30 syringe filters come prewashed, designed to minimize carbon leaching from the cellulose acetate membrane that could otherwise lead to sample contamination.
Specifications
- Catalog No:
- 514-0102
- 514-0288
- Rozměry:
- 26×34 mm
- 26×34 mm
- Maximální provozní tlak:
- 6,9 bar
- 6,9 bar
- Hmotnost:
- 4,7 g
- 4,7 g