Orders LinkContactUsLinkComponent
Flasks, round bottom, with two necks
Flasks, round bottom, with two necks
250 ml, 29/32 NS, 29/32 NS
Catalog # WITG0.754.250
Supplier:   witeg Labortechnik
Flasks, round bottom, with two necks
Catalog # WITG0.754.250
Supplier:  witeg Labortechnik
Flasks, round bottom, with two necks
Flasks, round bottom, with two necks
250 ml, 29/32 NS, 29/32 NS
Price
1 190,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Borosilicate glass 3.3 two necks, parallel side neck.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One vertical centre neck and one parallel side neck. With matt white labelling area.
Specifications
  • Kapacita:
    250 ml
  • Boční hrdla:
    29/32 NS
  • Centrální hrdlo:
    29/32 NS