ContactUsLinkComponent
Accessories for spectrophotometric colorimeters, Lovibond® model F
Accessories for spectrophotometric colorimeters, Lovibond® model F
Catalog # TINT180110
Dodavatel:  Lovibond Tintometer
Accessories for spectrophotometric colorimeters, Lovibond® model F
Catalog # TINT180110
Dodavatel:  Lovibond Tintometer
Specifikace
  • Popis:
    Replacement sample chamber insert for Model F
  • Sekvence:
    30
  • Bal.:
    3