Accessories for pH/°C meter, handheld
Supplier: Testo
Certificates
About this item
Frequently Bought Together
Ethylacetát ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
Supplier: VWR Chemicals
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® HCO 304, Konduktometr ruční
Supplier: VWR Collection
Accessories for microbiology manifold
Supplier: Cytiva (Formerly Pall Lab)
Accessories for pH/°C meter, handheld
Supplier: Testo
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Chlorid draselný 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Contact plates, RT/RTplus, single bagged
Supplier: MERCK LIFE SCIENCE
Monitory nápojů Microcheck® II
Supplier: Cytiva (Formerly Pall Lab)