Stands for Imhoff sedimentation cones or seperatory funnels, SP Bel-Art
Catalog # BELAH189670000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Určeno pro:eight cones or funnels
- Bal.:1
Specifications
About this item
This high-density polyethylene rack holds four separatory funnels on the lower shelf or four long-stem funnels on the upper shelf [maximum funnel diameter 98 mm (3⅞")].
- Set screws allow shelf height adjustments anywhere along the 74 cm (29") support rods
- Includes 24 insert clips that allow the rack to accommodate different sized Separatory Funnels and Imhoff Cones
- Chemical resistant and fully adjustable
Dimensions: 22×66×74 cm H (8,5×26×29")