Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer), Sigma-Aldrich®
Catalog # SIALT7777-1L
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:1 L
- Popis:TRIS-glycine-SDS buffer solution 10X concentrate (TGS buffer)
- M-Clarity quality level:MQ200