Pestle with extended handle, Bel-Art ProCulture, SP Bel-Art
Catalog # 212-7310
Supplier: Bel-Art Products, a Part of SP
Specifications
- Popis:For 1,5 ml microcentrifuge tubes, with rounded tip cone
- Length:210 mm
- Environmentally Preferable:
- Bal.:6
Specifications
About this item
Polypropylene tissue grinder efficiently homogenizes tissue and cells in microcentrifuge, centrifuge, and grinding tubes.
- Polypropylene construction
- Rounded end design
- Tapered conical tip
The cone tip tapers from 8,2 mm (0,33") down to 2,9 mm (0,12") in a length of 19,8 mm (0,78"). It features a rounded end. Overall length is 21 cm (8¼"), with a 4,7 mm (³/₁₆") diameter shaft. Steam autoclavable at 121 °C (250 °F).