Tubing connector assortment, SP Bel-Art
Catalog # 241-2271
Supplier: Bel-Art Products, a Part of SP
Specifications
- Bal.:1 SET
Specifications
About this item
Complete fitting assortment box with connectors for tubings.
- Assortment of 74 connector fittings
- Clear, hinge-top, polystyrene storage box
Ordering information: Kits consists of polypropylene tubing connectors 5 pairs, straight stepped tubing connectors 6 pairs, "T" connectors 5 pairs, "Y" connectors 6 pairs and polyethylene quick disconnects 3 pairs, cup-type 12 pairs.