Sampling knives, disposable, Sterileware®, SP Bel-Art
Catalog # 231-0016
Supplier: Bel-Art Products, a Part of SP
Specifications
- Type:Serrated and smooth edge blade
- Celková délka:209 mm
- Bal.:200
Specifications
About this item
PS, white.
- Ideal for food sampling, e.g. meats, vegetables, seafood and other fibrous and semi solid materials
- All knives are lot traceable
- Jagged blade cuts efficiently, opposite edge can spread material
- Individually packaged and sterilised
Caution: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904