0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Těsnění
Těsnící kroužky
Filter seal, Vac-Ring™, SP Bel-Art
Filter seal, Vac-Ring™, SP Bel-Art
BELAH389590000
:
Bel-Art Products, a Part of SP
Filter seal, Vac-Ring™, SP Bel-Art
BELAH389590000
:
Bel-Art Products, a Part of SP
Popis:
Filter seal, Vac-Ring™, black, for funnel stems up to 22 mm Ø, thickness 9,5 mm and O.Ø 76 mm
Materiál:
Neopren
Environmentally Preferable:
Bal.:
1