Magnetic stirring bar retriever, HDPE, SP Bel-Art
BELAF377750000
- Určeno pro:External use, avoids contact with the liquid and possible contamination
- Délka:200 mm
- Bal.:1
This retriever is able to remove a magnetic stirring bar without coming into contact with the contents of the vessel.
- Eliminates contamination of contents
- Does not need to be cleaned or autoclaved after use
- Long ribbed handle provides a secure grip