Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standardy
Standardy organické
Pentachlorobenzene 100 µg/ml in isooctane
Pentachlorobenzene 100 µg/ml in isooctane
Pentachlorobenzene 100 µg/ml in isooctane
Catalog # PROOCILULM72341.2
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
Pentachlorobenzene 100 µg/ml in isooctane
Catalog # PROOCILULM72341.2
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Specifications
Typ balení:
Glass bottle
Bal.:
1,2 mL
Popis:
Pentachlorobenzene 100 µg/ml in isooctane
Specifications
About this item
Detaily produktu
Detaily produktu