Food butyrometer, without stopper
Katalogové číslo 614-0135
Supplier: GERBER FUNKE
Specifications
- Popis:Butyrometer for food, Roeder´s, 0 - 100 % : 1,0
- Bal.:10
Specifications
About this item
According to Roeder´s weighing method.
According to Roeder´s weighing method.