Orders LinkContactUsLinkComponent
Dissection set
Dissection set
Dissection set in a wooden case, 9 pcs.
Katalogové číslo HERE1110401
Dodavatel:   HERENZ HEINZ
Dissection set
Katalogové číslo HERE1110401
Dodavatel:  HERENZ HEINZ
Dissection set
Dissection set
Dissection set in a wooden case, 9 pcs.
Přejít na:
Price
2 150,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
This dissection set includes nine instruments in a strong wooden case.
Content:
1 scalpel handle No. 4 with two blades
1 scalpel
1 scalpel handle No. 3 with two blades
1 forceps
1 scissors (pointed)
2 dissection needles (1 pointed, 1 with blade)
Specifikace
  • Popis:
    Dissection set in a wooden case, 9 pcs.