Mini tongs, Sterileware®, SP Bel-Art
Katalogové číslo BELAF379440000
Dodavatel: Bel-Art Products, a Part of SP
Specifikace
- Délka:110 mm
- Bal.:25
- Typ:Sterile
Specifikace
O tomto produktu
PS, white. For handling food, biological, chemical or pharmaceutical samples.
- Individually packaged, lot traceable and sterilized sample handling tongs
- Molded from smooth white polystyrene
- Serrated jaws securely hold small quantities of moist or dry material
- Disposable, designed for one time use to avoid cross contamination
Caution: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904