Desiccator plates, high heat, Minerit HD, SP Bel-Art
Catalog # BELAF420380230
Supplier: Bel-Art Products, a Part of SP
Specifications
- Ø:230 mm
- Type:Fibre reinforced cement board
- Sekvence:03
- Bal.:1
Specifications
About this item
Withstand temperatures up to 149 °C (300 °F). Contain no asbestos and are non combustible, stable and chemical resistant. Circular and perforated with 6 mm diameter holes on 25 mm centers in a straight line pattern.