Orders LinkContactUsLinkComponent
Storage Drawer
Storage Drawer
Durable waterproof drawer for long term storage, 405 × 230 × 51 mm
Catalog # 720-0283
Supplier:   Simport Scientific
Storage Drawer
Catalog # 720-0283
Supplier:  Simport Scientific
Storage Drawer
Storage Drawer
Durable waterproof drawer for long term storage, 405 × 230 × 51 mm
Price
4 060,00 Kč
Log in for availability
1 * 12 KS
Product Details & Documents
Durable, waterproof, heavy-duty cardboard.
  • Requires minimal space
  • Label provided for each box allows recording of specimen number, date and type of specimen
  • Each box can store up to 165 embedding rings or 250 cassettes
Specifications
  • Popis:
    Durable waterproof drawer for long term storage, 405 × 230 × 51 mm