Order Entry
Czech Republic
ContactUsLinkComponent
4-tert-Butylcalix[4]arene for synthesis, Sigma-Aldrich®
4-tert-Butylcalix[4]arene for synthesis, Sigma-Aldrich®
Catalog # 8.14707.0001
Supplier:  Merck
CAS Number:  
4-tert-Butylcalix[4]arene for synthesis, Sigma-Aldrich®
Catalog # 8.14707.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Typ balení:
    Glass bottle
  • Bal.:
    1 g
  • Sekvence:
    3047

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    344 - 347 °C
  • Molar Mass:
    648.92 g/mol
  • Hill Formula:
    C44H56O4

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
CAS Number:60705-62-6
Formula:C₄₄H₅₆O₄
MDL Number:MFCD00066280