trans,trans-2,4-Decadienal, Sigma-Aldrich®
Catalog # SIAL180513-5G
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:5 g
- Sekvence:3173
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
EINECS:246-668-9
CAS Number:25152-84-5
Formula:C₁₀H₁₆O
MDL Number:MFCD00007007