0
0,00 Kč
Order Entry
Přihlásit se
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Zadání objednávky
Stav objednávky
Váš účet
Košík
Czech Republic
Zadání objednávky
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Domů
Nábytek kancelářský
Police
Shelf for desiccator cabinets, SICCO Big Star
Shelf for desiccator cabinets, SICCO Big Star
Shelf for desiccator cabinets, SICCO Big Star
Catalog # 467-0113
Supplier:
Bohlender
Expand image
Shelf for desiccator cabinets, SICCO Big Star
Catalog # 467-0113
Supplier:
Bohlender
Specifications
Popis:
Stainless steel shelf with 2 aluminium rails
W×D×H:
515×495×0,8 mm
Environmentally Preferable:
Bal.:
1
For:
Big star and Maxi desiccators
Specifications
About this item
Product Details
Product Details