Orders LinkContactUsLinkComponent
Adapter for Tubes, PTFE
Adapter for Tubes, PTFE
Adapter for tube, PTFE; Filter Ø: 5 mm, ¹/₈" | 3 m/m
Katalogové číslo ROBU18510
Dodavatel:   ROBU GLASFILTER
Adapter for Tubes, PTFE
Katalogové číslo ROBU18510
Dodavatel:  ROBU GLASFILTER
Adapter for Tubes, PTFE
Adapter for Tubes, PTFE
Adapter for tube, PTFE; Filter Ø: 5 mm, ¹/₈" | 3 m/m
Přejít na:
Price
402,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
ROBU® Adapter for tubes made from PTFE provides a secure, leak-proof connection between tubing and equipment. Ideal for chemical, pharmaceutical, and laboratory applications, offering high chemical resistance and durability.
  • Made from durable PTFE
  • Resistant to chemicals and solvents
  • Leak-proof connection
  • Compatible with a wide range of tubes
  • High thermal stability
  • Ideal for laboratory and industrial use
  • Reusable and easy to install
ROBU Adapter for Tubes, crafted from high-quality PTFE (polytetrafluoroethylene), provides a secure and leak-proof connection for laboratory, pharmaceutical, and industrial applications. PTFE's exceptional chemical resistance ensures the adapter's compatibility with a wide range of chemicals and solvents, making it perfect for demanding environments.
The adapter’s excellent thermal stability allows it to perform reliably at high temperatures. It is easy to install, reusable, and ensures a tight seal for effective fluid or gas transfer, making it an essential component for various laboratory setups and industrial processes.
Documents
Specifikace
  • Hadice int.Ø:
    ¹/₈" | 3 m/m
  • Popis:
    Adapter for tube, PTFE; Filter Ø: 5 mm