Pipette stand for single channel pipettes, WHEATON®
Catalog # WHEAW851361
Dodavatel: DWK Life Sciences
Specifikace
- Určeno pro:7 single channel pipettes
- Barva:Světle šedá
- Popis:Pipettor universal workstation, model 337, with anti-slip pads
- Bal.:1
Specifikace
O tomto produktu
This space-saving pipette stand holds up to seven single channel pipettes of most brands.
- Made of sturdy polyamid material, easy to clean
- Anti-slip pads guarantee bench stability
- Work station fits new and old generations of Socorex and other brand pipettes