Single-channel pipettes, mechanical, fixed / variable volume, Proline® Plus
O tomto produktu
High quality pipettes with new, innovative design and technology, combining safety, ergonomics, efficiency in pipetting, with easy calibration and maintenance. They include built-in replaceable tip cone filters, which prevent pipette contamination and damage.
- Good ergonomics - handle design suitable for both left- and right-handed users, clear and easy to read display with big numbers and small increments, light and easy tip loading and ejection
- Fit the most commonly used lab tips thanks to slim tip ejector collar
- Plunger technology results in the low pipetting force
- Colour-coded volume range for easy pipette selection
- Good chemical and UV resistance; thermal insulation of the internal components improves both accuracy and reproducibility
- Fully autoclavable
There are two types of filter available: Safe-Cone Standard filter and Safe-Cone Plus filter. It is recommended to use the Standard filter for general applications and the Plus filter for more demanding applications, such as cell culture, bacterial, viral work and molecular biology.
* Safe-Cone Plus.
Starter Kit 1 includes two Proline® Plus pipettes (0,1 - 3 µl / 0,5 - 10 µl), two pipette holders and one calibration tool for each pipette, two single-tray tip boxes, literature.
Starter Kit 2 includes three Proline® Plus pipettes (0,5 - 10 µl / 10 - 100 µl / 100 - 1000 µl), two pipette holders and one calibration tool for each pipette, three single-tray tip boxes, literature.
Starter Kit 3 includes three Proline® Plus pipettes (2 - 20 µl / 20 - 200 µl / 100 - 1000 µl), two pipette holders and one calibration tool for each pipette, three single-tray tip boxes, literature.
: Proline® Plus single pipettes are supplied with calibration tool, autoclavable grease, tip, Safe-Cone filters (>10 µl), forceps, user manual and QC certificate.
Starter Kits are also available.
21 Dostupných variant níže
Product Details & Documents
High quality pipettes with new, innovative design and technology, combining safety, ergonomics, efficiency in pipetting, with easy calibration and maintenance. They include built-in replaceable tip cone filters, which prevent pipette contamination and damage.
- Good ergonomics - handle design suitable for both left- and right-handed users, clear and easy to read display with big numbers and small increments, light and easy tip loading and ejection
- Fit the most commonly used lab tips thanks to slim tip ejector collar
- Plunger technology results in the low pipetting force
- Colour-coded volume range for easy pipette selection
- Good chemical and UV resistance; thermal insulation of the internal components improves both accuracy and reproducibility
- Fully autoclavable
There are two types of filter available: Safe-Cone Standard filter and Safe-Cone Plus filter. It is recommended to use the Standard filter for general applications and the Plus filter for more demanding applications, such as cell culture, bacterial, viral work and molecular biology.
* Safe-Cone Plus.
Starter Kit 1 includes two Proline® Plus pipettes (0,1 - 3 µl / 0,5 - 10 µl), two pipette holders and one calibration tool for each pipette, two single-tray tip boxes, literature.
Starter Kit 2 includes three Proline® Plus pipettes (0,5 - 10 µl / 10 - 100 µl / 100 - 1000 µl), two pipette holders and one calibration tool for each pipette, three single-tray tip boxes, literature.
Starter Kit 3 includes three Proline® Plus pipettes (2 - 20 µl / 20 - 200 µl / 100 - 1000 µl), two pipette holders and one calibration tool for each pipette, three single-tray tip boxes, literature.
: Proline® Plus single pipettes are supplied with calibration tool, autoclavable grease, tip, Safe-Cone filters (>10 µl), forceps, user manual and QC certificate.
Starter Kits are also available.
Specifications for Product Family
Specifikace
- Catalog No:
- 613-5160
- 613-5161
- 613-5162
- 613-1753
- 613-5163
- 613-5164
- 613-5165
- 613-5166
- 613-5167
- 613-5168
- 613-5169
- 613-5170
- 613-5171
- 613-5172
- 613-5173
- 613-5174
- 613-5175
- 613-5176
- 613-5177
- 613-5178
- 613-5179
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certifikát ochrany životního prostředí:
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
- Renewable energy certificate; ISO 14001
Specifikace
Product Family Options
Variable volume
Informace o produktu
- SekvenceObjemIncrementPřesnostSprávnostPro pipetovací špičkyS bezpečným-kónickým filtrem (y)DostupnostPrice
- 010,3 - 3 µl0,002 µl±1,30 - ±10,00 %<0,80 - <6,00 %10 µl—Přihlásit se pro dostupnost1 * 1 KS6 830,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5160Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - 021 - 10 µl0,01 µl±1,00 - ±5,00 %<0,60 - <4,00 %10 µl—Přihlásit se pro dostupnost1 * 1 KS6 830,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5161Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - 032 - 20 µl0,02 µl±0,90 - ±3,00 %<0,40 - <2,00 %300 µl613-4072Přihlásit se pro dostupnost1 * 1 KS6 830,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5162Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - 045 - 50 µl0,10 µl±0,90 - ±3,00 %<0,30 - <1,60 %300, 350 µl613-4071, 613-0694*Přihlásit se pro dostupnost1 * 1 KS6 830,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-1753Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - 0510 - 100 µl0,10 µl±0,80 - ±2,00 %<0,15 - <1,00 %300, 350 µl613-4071, 613-0694*Přihlásit se pro dostupnost1 * 1 KS6 830,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5163Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - 0620 - 200 µl0,20 µl±0,60 - ±2,00 %<0,15 - <0,80 %300, 350 µl613-4070, 613-0693*Přihlásit se pro dostupnost1 * 1 KS6 830,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5164Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - 07100 - 1000 µl1,00 µl±0,60 - ±1,00 %<0,20 - <0,40 %1000 µl613-4069, 613-4074*Přihlásit se pro dostupnost1 * 1 KS6 830,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5165Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - 08500 - 5000 µl10,00 µl±0,50 - ±2,00 %<0,20 - <0,60 %5000 µl613-4068, 613-0692*Přihlásit se pro dostupnost1 * 1 KS6 830,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5166Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - 091000 - 10000 µl20,00 µl±0,60 - ±3,00 %<0,20 - <0,60 %10000 µl613-4068, 613-0692*Přihlásit se pro dostupnost1 * 1 KS6 830,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5167Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact
Fixed volume
Informace o produktu
- SekvenceObjemIncrementPřesnostSprávnostPro pipetovací špičkyS bezpečným-kónickým filtrem (y)DostupnostPrice
- Specifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5168Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - Specifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5169Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - Specifikace:
More
Specifications
Cat. No.613-5170Environmentally PreferableCertifikát ochrany životního prostředí - Specifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5171Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - Specifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5172Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - 15100 µl±0,50 %<0,30 %300, 350 µl613-4071, 613-0694*Přihlásit se pro dostupnost1 * 1 KS4 600,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5173Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - 16200 µl±0,40 %<0,20 %300, 350 µl613-4070, 613-0693*Přihlásit se pro dostupnost1 * 1 KS4 600,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5174Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - Specifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5175Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - Specifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5176Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - Specifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5177Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - Specifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5178Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact - Specifikace:
Low Manufacturing Impact More
Specifications
Cat. No.613-5179Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable energy certificate; ISO 14001Low Manufacturing Impact
Multi-packs
Informace o produktu
- Katalogové čísloSekvencePopisDostupnostPrice
- 043+1-pack, incl. three single channel pipettes (0,5 - 10 µl, 20 - 200 µl, 100 - 1000 µl), one 8-channel pipette (30 - 300 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 1× 0,5 - 200 µl, 1× 5 - 350 µl, 1× 10 - 1000 µl) and a linear standPřihlásit se pro dostupnost1 * 1 MJ32 750,00 Kč
Stands
Informace o produktu
- Katalogové čísloSekvenceBarvaPopisŠ×H×VDostupnostPrice
- 59Bílá/černáUniverzální stojan pro 11 pipet ve svislé poloze290×250×60 mmPřihlásit se pro dostupnost1 * 1 KS2 120,00 Kč
Accessories
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Safe-Cone filters, Standard, Ø 1,83 mmSafe-Cone filters, Standard, Ø 1,83 mm613-407250Přihlásit se pro dostupnost1 * 50 KS - 880,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Safe-Cone filters, Plus, Ø 6,73 mm
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Flitry Safe-Cone, Standard, Ø 3,15 mm
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Safe-Cone filters, Standard, Ø 6,73 mmSafe-Cone filters, Standard, Ø 6,73 mm613-406854Přihlásit se pro dostupnost1 * 50 KS - 880,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Flitry Safe-Cone, Standard, Ø 2,51 mm
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Flitry Safe-Cone, Standard, Ø 5,33 mm
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Flitry Safe-Cone, Plus, Ø 5,33 mm
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Flitry Safe-Cone, Plus, Ø 2,51 mm
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Flitry Safe-Cone, Plus, Ø 3,15 mm