ContactUsLinkComponent
Viewing cabinets, Chromato-Vue®, C-75G
Viewing cabinets, Chromato-Vue®, C-75G
  UVPA95-0253-02
undefined
Viewing cabinets, Chromato-Vue®, C-75G
  UVPA95-0253-02
  • Model:
    C-75
  • Vlnová délka:
    254/365 nm
  • Popis:
    Chromato-Vue® C-75, high performance, with two UV tubes
  • Výstup:
    2×15 W
  • Bal.:
    1
  • Určeno pro:
    Fluorescence studies and non destructive testing