Přeskočit na Patička
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Vzorkovače
Vzorkovače kapalné
Accessories for E-Z Sampler®
Accessories for E-Z Sampler®
Accessories for E-Z Sampler®
Catalog # WHEA885025
Supplier:
DWK Life Sciences
Expand image
Accessories for E-Z Sampler®
Catalog # WHEA885025
Supplier:
DWK Life Sciences
Specifications
Popis:
Bottles with lined cap, 12 ml
Sekvence:
901
Bal.:
12
Specifications
About this item
Product Details
Product Details