Scrubbing sponge for glassware, Cleanware™, SP Bel-Art
Catalog # BELAF170780001
Supplier: Bel-Art Products, a Part of SP
Specifications
- Balené:2 sponges/pack
- Barva:Žlutá/zelená
- D׊:108×64 mm
- Tloušťka:25 mm
- Bal.:2
Specifications
About this item
This odor-resistant, highly absorbent sponge is designed to remove stubborn residue on glassware.
- Wipe dry performance equal to cellulose; maintains the desirable physical properties of polyurethane
- Foam contains anti-microbial technology to inhibit odor causing bacteria within the sponge itself
- Stays soft and flexible, wet or dry; will not crack or shrink, or become brittle like cellulose
- Scouring side resists snagging; does not leave fibers behind and retains aggressive scouring ability even with frequent use