Dispenser, Measure-Matic™, SP Bel-Art
Catalog # BELAF116540000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Objem:5 - 25 ml
- Bal.:1
Specifications
About this item
Bottles, LDPE, translucent, for quick dispensing, with polypropylene measuring cup. Measuring cup slides onto a polypropylene delivery tube. The height of the tube determines the volume of liquid dispensed into the measuring cup when the bottle is squeezed.
- Repetitive dispensing of the same volume of liquid
- When the bottle pressure is released excess flluid returns into the bottle for spill free pouring
- Good chemical resistance