ContactUsLinkComponent
qPCR Mycoplasma Test Kit
qPCR Mycoplasma Test Kit
Catalog # A9019.0025
Supplier:  PanReac AppliChem
CAS Number:  
undefined
qPCR Mycoplasma Test Kit
Catalog # A9019.0025
Supplier:  PanReac AppliChem
CAS Number:  
Specifications
  • Bal.:
    25 Tests
  • Popis:
    Testovací sada qPCR mycoplasma
Specifications
About this item
Lyophilised PCR mix for the detection of mycoplasma in cell culture by Real-Time/qPCR.
Delivery: Supplied with Taq DNA polymerase. Kit contains PCR primer mix (incl. primers, probes, nucleotides), reaction buffer solution, water (PCR grade), positive template control, and internal control DNA.