Infusion filter, Sterifix® Paed
720-2580
: B. Braun
- Barva:Bezbarvá
- Popis:Infusion filters
- Bal.:50
Sterifix Paed is specially designed for the bacteria and particle retention.
- Reliable air venting regardless of position
- Flow rate (aqua dest.) over 10 ml per minute
- Luer Lock fitting