Biocide, sterile filtered, Klercide™ Sporicidal Low Residue Peroxide
Supplier: ECOLAB CONTAMINATION CONTROL
Certificates
About this item
Suitable for ISO Class 5, Grade A and B (microbial count) areas: cleanroom processed and sterile, double bagged. Blend of 6% hydrogen peroxide and deionised water, with broad spectrum activity including bacterial spores. Filtered to 0,2 µm.
- CEN and isolate tested
- Safe and effective; non toxic and non corrosive
- Fully compatible and ideal for a rotational regime
- SteriShield Delivery System™ ensures 3 months in-use shelf life
Certifications: EN standards: EN 1276 - bactericidal activity, EN 1650 - fungicidal activity, EN 13697 - bactericidal surface test, EN 13704 - sporocidal activity, EN 14476 - virucidal.
Caution: Use biocides safely. Always read the label and product information before use.
Frequently Bought Together