Sampling knives, sterile, Corning® Gosselin™
231-0058
: Corning
- Barva:Bílá
- Popis:Sampling tools, knives, sterile
- Celková délka:175 mm
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:RECs, ISO 14001 - Borre
- Bal.:1.000
PS, white. Knife for taking samples, as well as a variety of other uses.
- For the removal of small, solid samples
- Irradiated, traceable by the printed expiry date on the bottom of the packaging
- Ideal for single use
: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904