Baskets for test tubes
Basket for washing and storing test tubes, aluminium, M, 256×152×152 mm
Katalogové číslo HEATHS20341B
Dodavatel: HEATHROW
Přejít na:
Product Details & Documents
These test tube baskets are made of aluminium.
- Lightweight
- Autoclavable
- Sturdy
They are designed for rinsing, transporting and storing laboratory glassware.
Specifikace
- Popis:Basket for washing and storing test tubes, aluminium, M
- Š×H×V:256×152×152 mm