0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Standardy
Standardy organické
Triclosan 10 µg/ml in methanol
Triclosan 10 µg/ml in methanol
PROONE9337
:
LGC Standards PROMOCHEM
:
Triclosan 10 µg/ml in methanol
PROONE9337
:
LGC Standards PROMOCHEM
:
Typ balení:
Glass bottle
Bal.:
10 mL
Čí. CAS:
3380-34-5
Popis:
Triclosan 10 µg/ml in methanol
Vzorec:
C₁₂H₇Cl₃O₂
Popis:
Triclosan