Order Entry
Czech Republic
ContactUsLinkComponent
D-Panthenol 10 µg/ml in methanol
D-Panthenol 10 µg/ml in methanol
Catalog # PROONE9298
CAS Number:  
D-Panthenol 10 µg/ml in methanol
Catalog # PROONE9298
CAS Number:  

Specifications

  • Typ balení:
    Glass bottle
  • Bal.:
    10 mL
  • Čí. CAS:
    81-13-0
  • Popis:
    D-Panthenol 10 µg/ml in methanol
  • Vzorec:
    C₉H₁₉NO₄
  • Popis:
    D-Panthenol

Specifications

About this item