Cleanroom gloves, latex, BioClean Legacy™
Supplier: Nitritex
Certificates
About this item
These ambidextrous cleanroom gloves with beaded cuffs can be used in medical as well as in laboratory environments. Suitable for double-donning.
- Protein level less than 50 µg/g
- Textured finish for improved grip
- Latex provides exceptional flexibility and comfort
- Beaded cuff provides easier donning and stability on the arm
- Each size carries a unique barcode on the carton (ITF-14), which allows automatic data entry into stock control systems
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
EN 455, EN 374, Cat. III; AQL 1.5
Certifications: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Suitable for contact with dry, alcoholic and acidic foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Suitable for contact with dry, alcoholic and acidic foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Packaging: 100 pieces per sealed inner PE bag; one sealed inner PE bag per sealed outer PE bag; 10 outer PE bags per carton liner; one carton liner per carton (1000 pieces).
Specifications
- Catalog No:
- 112-3308
- 112-3304
- 112-3305
- 112-3306
- 112-3307
- 112-3303
- Obouruké/jednoruké:
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Jednorázové/znovupoužitelné:
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Pudrované/nepudrované:
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Třída čistoty:
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- Typ manžety:
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Tloušťka límce:
- 0,11 mm
- 0,11 mm
- 0,11 mm
- 0,11 mm
- 0,11 mm
- 0,11 mm
- Tloušťka prstu:
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- Tloušťka drapáku:
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- Norma EN:
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374